Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032947.1 | NP_001028119.1 | 168 | 79 | ||
GenBank mRNA | AF449285.1 | - | 168 | 79 | ||
JU492267.1 | - | 70 | 79 | |||
JV694538.1 | - | 148 | 79 | |||
KT359585.1 | - | 203 | 79 |
Gene Symbol: LOC100423131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028850129.1 | XP_028705962.1 | 900 | 79 | ||
XM_028850130.1 | XP_028705963.1 | 1145 | 79 | |||
XM_028850131.1 | XP_028705964.1 | 1052 | 79 | |||
XM_028850132.1 | XP_028705965.1 | 1206 | 79 | |||
XM_028850133.1 | XP_028705966.1 | 961 | 79 | |||
XM_028850134.1 | XP_028705967.1 | 779 | 79 | |||
XM_028850136.1 | XP_028705969.1 | 1024 | 79 | |||
XM_028850137.1 | XP_028705970.1 | 722 | 79 | |||
GenBank mRNA | JV453160.1 | - | 186 | 79 | ||
JV554243.1 | - | 182 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta Inventoried |