Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047137.1 | NP_001040602.1 | 356 | 78 | ||
GenBank mRNA | AY349163.1 | - | 356 | 78 | ||
AY445834.1 | - | 356 | 78 | |||
JV045380.1 | - | 354 | 78 | |||
JV669635.1 | - | 409 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |