Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032911.1 | NP_001028083.1 | 2114 | 68 | ||
GenBank mRNA | AY526325.1 | - | 2114 | 68 | ||
JV650576.1 | - | 357 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |