Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033020.1 | NP_001028192.1 | 1320 | 79 | ||
XM_015126171.2 | XP_014981657.1 | 1913 | 79 | |||
XM_015126172.2 | XP_014981658.1 | 1898 | 79 | |||
XM_028840347.1 | XP_028696180.1 | 1418 | 79 | |||
GenBank mRNA | AY845352.1 | - | 1320 | 79 | ||
JU323293.1 | - | 1554 | 79 | |||
JU470897.1 | - | 1564 | 79 | |||
JU470898.1 | - | 1549 | 79 | |||
JV601061.1 | - | 1554 | 79 | |||
JV635411.1 | - | 1528 | 79 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Macaca mulatta |