Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042638.2 | NP_001036103.2 | 71 | 70 | ||
XM_028832634.1 | XP_028688467.1 | 274 | 70 | |||
GenBank mRNA | CN647022.1 | - | 71 | 70 | ||
JU321360.1 | - | 171 | 70 | |||
JU473604.1 | - | 43 | 70 | |||
JV048086.1 | - | 59 | 70 | |||
U93310.2 | - | 31 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |