Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUCKS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266481.1 | NP_001253410.1 | 462 | 158 | ||
GenBank mRNA | JU319617.1 | - | 470 | 158 | ||
JU319618.1 | - | 470 | 158 | |||
JU471457.1 | - | 575 | 158 | |||
JV625906.1 | - | 622 | 158 | |||
JV635557.1 | - | 462 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |