Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUCKS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266481.1 | NP_001253410.1 | 607 | 63 | ||
XM_015118335.2 | XP_014973821.1 | 780 | 63 | |||
GenBank mRNA | JU319617.1 | - | 615 | 63 | ||
JU319618.1 | - | 615 | 63 | |||
JU471457.1 | - | 720 | 63 | |||
JV625906.1 | - | 767 | 63 | |||
JV635557.1 | - | 607 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |