Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRAID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193926.2 | NP_001180855.1 | 601 | 79 | ||
XM_028831453.1 | XP_028687286.1 | 631 | 79 | |||
XM_028831454.1 | XP_028687287.1 | 387 | 79 | |||
GenBank mRNA | JU334872.1 | - | 614 | 79 | ||
JU487868.1 | - | 433 | 79 | |||
JV043224.1 | - | 568 | 79 |
Gene Symbol: SLC5A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV664632.1 | - | 640 | 79 |