Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFB1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261482.1 | NP_001248411.1 | 165 | 73 | ||
XM_028846716.1 | XP_028702549.1 | 227 | 73 | |||
XM_028846717.1 | XP_028702550.1 | 227 | 73 | |||
XM_028846718.1 | XP_028702551.1 | 227 | 73 | |||
XM_028846719.1 | XP_028702552.1 | 214 | 73 | |||
XM_028846720.1 | XP_028702553.1 | 221 | 73 | |||
GenBank mRNA | JU334782.1 | - | 165 | 73 | ||
JV685836.1 | - | 198 | 73 |
Gene Symbol: TIAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV704029.1 | - | 3666 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |