Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266410.1 | NP_001253339.1 | 109 | 124 | ||
XM_015131604.2 | XP_014987090.1 | 102 | 124 | |||
GenBank mRNA | JU332745.1 | - | 109 | 124 | ||
JU473132.1 | - | 109 | 124 | |||
JV046074.1 | - | 109 | 124 | |||
JV601896.1 | - | 136 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |