Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261368.1 | NP_001248297.1 | 300 | 101 | ||
GenBank mRNA | JU322518.1 | - | 262 | 101 | ||
JU473306.1 | - | 326 | 101 | |||
JV045674.1 | - | 300 | 101 | |||
JV601919.1 | - | 344 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |