Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104583.1 | NP_001098053.1 | 454 | 93 | ||
GenBank mRNA | EF057492.1 | - | 454 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |