Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPM6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265765.1 | NP_001252694.1 | 1039 | 65 | ||
XM_015139515.2 | XP_014995001.1 | 753 | 65 | |||
XM_015139516.2 | XP_014995002.1 | 703 | 65 | |||
XM_015139517.2 | XP_014995003.1 | 981 | 65 | |||
XM_028848000.1 | XP_028703833.1 | 366 | 65 | |||
XM_028848001.1 | XP_028703834.1 | 366 | 65 | |||
GenBank mRNA | JV667166.1 | - | 1039 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |