Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257642.2 | NP_001244571.1 | 636 | 127 | ||
XM_028830388.1 | XP_028686221.1 | 957 | 127 | |||
GenBank mRNA | JU337033.1 | - | 637 | 127 | ||
JU473225.1 | - | 934 | 127 | |||
JV044588.1 | - | 795 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |