Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261087.1 | NP_001248016.1 | 736 | 71 | ||
GenBank mRNA | JU328560.1 | - | 294 | 71 | ||
JU477636.1 | - | 774 | 71 | |||
JV048570.1 | - | 736 | 71 | |||
JV664397.1 | - | 860 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |