Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU492877.1 | - | 67 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |