Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265876.1 | NP_001252805.1 | 174 | 153 | ||
GenBank mRNA | JU334545.1 | - | 179 | 153 | ||
JU475631.1 | - | 188 | 153 | |||
JV047082.1 | - | 174 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |