Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257433.1 | NP_001244362.1 | 121 | 116 | ||
XM_015125574.2 | XP_014981060.1 | 1372 | 116 | |||
GenBank mRNA | JU321035.1 | - | 121 | 116 | ||
JU476037.1 | - | 116 | ||||
JV045175.1 | - | 116 | ||||
JV653914.1 | - | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |