Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265898.1 | NP_001252827.1 | 305 | 62 | ||
XM_015132342.2 | XP_014987828.1 | 439 | 62 | |||
GenBank mRNA | JU333753.1 | - | 354 | 62 | ||
JU474994.1 | - | 305 | 62 | |||
JU474995.1 | - | 305 | 62 | |||
JU474996.1 | - | 305 | 62 | |||
JU474997.1 | - | 305 | 62 | |||
JV668067.1 | - | 386 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |