Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1C4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001118631.4 | XP_001118631.3 | 150 | 68 | ||
XM_028825937.1 | XP_028681770.1 | 131 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |