Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261315.1 | NP_001248244.1 | 831 | 96 | ||
XM_028852179.1 | XP_028708012.1 | 1109 | 96 | |||
XM_028852180.1 | XP_028708013.1 | 278 | 96 | |||
GenBank mRNA | JU319897.1 | - | 811 | 96 | ||
JU470826.1 | - | 819 | 96 | |||
JV044223.1 | - | 831 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |