Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320503.1 | NP_001307432.1 | 566 | 75 | ||
NM_001320504.1 | NP_001307433.1 | 566 | 75 | |||
XM_015114275.2 | XP_014969761.1 | 590 | 75 | |||
XM_028832689.1 | XP_028688522.1 | 527 | 75 | |||
XM_028832690.1 | XP_028688523.1 | 617 | 75 | |||
GenBank mRNA | JU322036.1 | - | 533 | 75 | ||
JU473103.1 | - | 566 | 75 | |||
JV043856.1 | - | 496 | 75 | |||
JV463154.1 | - | 417 | 75 | |||
JV566967.1 | - | 549 | 75 | |||
JV602021.1 | - | 488 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |