Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257521.1 | NP_001244450.1 | 140 | 75 | ||
GenBank mRNA | JU320026.1 | - | 140 | 75 | ||
JU471759.1 | - | 126 | 75 | |||
JV045724.1 | - | 140 | 75 | |||
JV566878.1 | - | 170 | 75 | |||
JV655073.1 | - | 116 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |