Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS2R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015139731.2 | XP_014995217.1 | 1206 | 128 | ||
XM_015139732.2 | XP_014995218.1 | 1719 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |