Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261200.1 | NP_001248129.1 | 524 | 77 | ||
XM_028843215.1 | XP_028699048.1 | 429 | 77 | |||
GenBank mRNA | JU334567.1 | - | 512 | 77 | ||
JV047102.1 | - | 524 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |