Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015130437.2 | XP_014985923.1 | 1422 | 118 | ||
XM_015130439.2 | XP_014985925.1 | 1181 | 118 | |||
GenBank mRNA | JU494926.1 | - | 696 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |