Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100208.3 | XP_001100208.1 | 288 | 180 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |