Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SALL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122227.2 | XP_014977713.2 | 3503 | 67 | ||
GenBank mRNA | JU327279.1 | - | 340 | 67 | ||
JV712165.1 | - | 150 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |