Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNRF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083609.4 | XP_001083609.2 | 298 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |