Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015114575.2 | XP_014970061.1 | 950 | 144 | ||
XM_015114576.2 | XP_014970062.1 | 950 | 144 | |||
XM_015114577.2 | XP_014970063.1 | 860 | 144 | |||
XM_028833532.1 | XP_028689365.1 | 403 | 144 | |||
XM_028833533.1 | XP_028689366.1 | 403 | 144 | |||
XM_028833534.1 | XP_028689367.1 | 408 | 144 | |||
XM_028833535.1 | XP_028689368.1 | 144 | 144 | |||
GenBank mRNA | JV048171.1 | - | 846 | 144 | ||
JV048172.1 | - | 846 | 144 | |||
JV048173.1 | - | 846 | 144 | |||
JV660606.1 | - | 942 | 144 | |||
JV660607.1 | - | 942 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |