Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HFE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260576.2 | NP_001247505.1 | 349 | 122 | ||
GenBank mRNA | JU334244.1 | - | 349 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |