Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107647.4 | XP_001107647.1 | 1338 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta Inventoried |