Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN28B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137022.2 | XP_014992508.1 | 699 | 103 | ||
GenBank mRNA | JV060743.1 | - | 143 | 103 | ||
JV651097.1 | - | 348 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |