Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPNMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266650.1 | NP_001253579.1 | 443 | 138 | ||
XM_015133861.2 | XP_014989347.1 | 596 | 138 | |||
XM_015133862.2 | XP_014989348.1 | 685 | 138 | |||
XM_015133863.2 | XP_014989349.1 | 597 | 138 | |||
XM_015133864.2 | XP_014989350.1 | 386 | 138 | |||
XM_015133865.2 | XP_014989351.1 | 665 | 138 | |||
GenBank mRNA | JU332515.1 | - | 463 | 138 | ||
JU473990.1 | - | 443 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |