Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028836908.1 | XP_028692741.1 | 2035 | 149 | ||
XM_028836909.1 | XP_028692742.1 | 2032 | 149 | |||
XM_028836910.1 | XP_028692743.1 | 2036 | 149 | |||
XM_028836912.1 | XP_028692745.1 | 2029 | 149 | |||
XM_028836913.1 | XP_028692746.1 | 2036 | 149 | |||
XM_028836914.1 | XP_028692747.1 | 2035 | 149 | |||
XM_028836915.1 | XP_028692748.1 | 2036 | 149 | |||
GenBank mRNA | JU474774.1 | - | 1943 | 149 | ||
JU474775.1 | - | 1943 | 149 | |||
JV609571.1 | - | 1941 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |