Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194155.1 | NP_001181084.1 | 366 | 68 | ||
XM_002806154.3 | XP_002806200.1 | 399 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |