Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194317.2 | NP_001181246.2 | 708 | 127 | ||
XM_015124040.2 | XP_014979526.2 | 1038 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |