Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMIGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194531.2 | NP_001181460.1 | 703 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |