Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028841368.1 | XP_028697201.1 | 1985 | 140 | ||
XM_028841369.1 | XP_028697202.1 | 1973 | 140 | |||
XM_028841371.1 | XP_028697204.1 | 1952 | 140 | |||
XM_028841372.1 | XP_028697205.1 | 1928 | 140 | |||
XM_028841373.1 | XP_028697206.1 | 2006 | 140 | |||
XM_028841374.1 | XP_028697207.1 | 2006 | 140 | |||
XM_028841375.1 | XP_028697208.1 | 1892 | 140 | |||
XM_028841376.1 | XP_028697209.1 | 1892 | 140 | |||
XM_028841377.1 | XP_028697210.1 | 1787 | 140 | |||
XM_028841378.1 | XP_028697211.1 | 2047 | 140 | |||
XM_028841379.1 | XP_028697212.1 | 1652 | 140 | |||
XM_028841380.1 | XP_028697213.1 | 1831 | 140 | |||
XM_028841381.1 | XP_028697214.1 | 1803 | 140 | |||
XM_028841383.1 | XP_028697216.1 | 1673 | 140 | |||
GenBank mRNA | JV706678.1 | - | 635 | 140 | ||
JV706679.1 | - | 635 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |