Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT6B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001108157.4 | XP_001108157.3 | 541 | 160 | ||
XM_028831757.1 | XP_028687590.1 | 158 | 160 | |||
XM_028831758.1 | XP_028687591.1 | 181 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |