Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015132387.2 | XP_014987873.2 | 1818 | 105 | ||
GenBank mRNA | JV051898.1 | - | 312 | 105 | ||
JV642405.1 | - | 292 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |