Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261335.1 | NP_001248264.1 | 50 | 98 | ||
XM_015135553.2 | XP_014991039.1 | 388 | 98 | |||
GenBank mRNA | JU337113.1 | - | 105 | 98 | ||
JU473281.1 | - | 50 | 98 | |||
JV633613.1 | - | 207 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta Inventoried |