Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194687.2 | NP_001181616.1 | 242 | 107 | ||
XM_001115537.4 | XP_001115537.2 | 394 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |