Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265962.1 | NP_001252891.1 | 293 | 92 | ||
XM_015132890.2 | XP_014988376.1 | 286 | 92 | |||
XM_015132891.2 | XP_014988377.1 | 336 | 92 | |||
XM_028844818.1 | XP_028700651.1 | 540 | 92 | |||
GenBank mRNA | JU471230.1 | - | 293 | 92 | ||
JU471231.1 | - | 293 | 92 | |||
JU471232.1 | - | 293 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |