Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136681.2 | XP_014992167.1 | 1856 | 72 | ||
XM_015136688.2 | XP_014992174.1 | 1821 | 72 | |||
XM_028835304.1 | XP_028691137.1 | 1442 | 72 | |||
GenBank mRNA | JU322260.1 | - | 1390 | 72 | ||
JU474436.1 | - | 1412 | 72 | |||
JV046270.1 | - | 1441 | 72 | |||
JV643321.1 | - | 1213 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |