Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257792.1 | NP_001244721.1 | 855 | 114 | ||
GenBank mRNA | JU320573.1 | - | 855 | 114 | ||
JU470962.1 | - | 854 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |