Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257468.1 | NP_001244397.1 | 1409 | 95 | ||
GenBank mRNA | JU321005.1 | - | 1409 | 95 | ||
JU473336.1 | - | 1407 | 95 | |||
JV044425.1 | - | 1321 | 95 |