Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194668.1 | NP_001181597.1 | 686 | 117 | ||
XM_015151001.2 | XP_015006487.1 | 736 | 117 | |||
GenBank mRNA | JV716030.1 | - | 634 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta Inventoried |