Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257817.1 | NP_001244746.1 | 72 | 101 | ||
XM_015119478.2 | XP_014974964.1 | 118 | 101 | |||
GenBank mRNA | JU322295.1 | - | 72 | 101 | ||
JU473672.1 | - | 73 | 101 | |||
JV602348.1 | - | 79 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |