Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257853.2 | NP_001244782.1 | 1601 | 146 | ||
XM_028835469.1 | XP_028691302.1 | 1702 | 146 | |||
XM_028835470.1 | XP_028691303.1 | 1711 | 146 | |||
XM_028835471.1 | XP_028691304.1 | 1741 | 146 | |||
GenBank mRNA | JU320853.1 | - | 1602 | 146 | ||
JU476916.1 | - | 1613 | 146 | |||
JV045337.1 | - | 1594 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |